Learn More
Sino Biological Recombinant Anti-CD5 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 50403R249A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD5 (rM CD5; Catalog#50403-M08H; NP_031676.3; Met1-Pro371) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-0348