Learn More
Bon Opus Biosciences Recombinant Mouse CD54/ICAM-1 (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP103850UG
Encompass_Preferred
Recombinant Mouse CD54/ICAM-1 (C-mFc) is produced by Human Cells. The target gene encoding Gln28-Asn485 is expressed with a C-mFc tag. Accession# AAH08626.1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1038
Catalog No. 50-217-2637