Learn More
Sino Biological Mouse CD55/DAF Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000786647
Encompass_Preferred
The Mouse CD55/DAF ELISA Pair Set is for the quantitative determination of Mouse CD55/DAF.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse CD55/DAF coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse CD55/DAF present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse CD55/DAF polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse CD55/DAF present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86647 Supplier Sino Biological Supplier No. SEK504685
The Mouse CD55/DAF ELISA Pair Set is for the quantitative determination of Mouse CD55/DAF.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse CD55/DAF coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse CD55/DAF present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse CD55/DAF polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse CD55/DAF present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.