Learn More
Sino Biological Recombinant Anti-CD55/DAF Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 50468R076A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD55 / DAF (rM CD55 / DAF; Catalog#50468-M08H; Q61475; Met1-Thr361) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-2510