Learn More
Sino Biological Mouse CD55 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 50468R07620
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD55 / DAF (rM CD55 / DAF; Catalog#50468-M08H; Q61475; Met1-Thr361).
Catalog No. 50-237-3139