Learn More
Sino Biological Mouse CD55 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 50110R133100
Encompass_Preferred
A DNA sequence encoding the Mouse CD55 (Q61475) (Met1-Thr361) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-4324