Learn More
BIOHIPPO Mouse CD59 Microsample
SDP
Catalog No. 5000315561
Encompass
Mouse CD59 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD59 in Serum plasma urine and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 11 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CD59 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CD59 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CD59 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15561 Supplier BIOHIPPO Supplier No. BHE15212536
Mouse CD59 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD59 in Serum plasma urine and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 11 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CD59 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CD59 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CD59 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color