Learn More
Sino Biological Recombinant Anti-CD59a Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50724R133100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD59a / Protectin (rM CD59a / Protectin; Catalog#50724-M08H; NP_001104530.1; Met1-Lys95).
Catalog No. 50-255-3122