Learn More
Sino Biological Recombinant Anti-CD59a Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50724R108P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD59a (rM CD59a; Catalog#50724-M08H; NP_001104530.1; Met1-Lys95) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-3123