Learn More
Sino Biological Recombinant Anti-CD5L Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50020R05420
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD5L (rM CD5L; Catalog#50020-M08H; NP_033820.2; Met1-Val352).
Catalog No. 50-241-1539