Learn More
BIOHIPPO Mouse CD5L Microsample
SDP
Catalog No. 5000313721
Encompass
Mouse CD5L Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD5L in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 14 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CD5L Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CD5L Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CD5L biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13721 Supplier BIOHIPPO Supplier No. BHE15210740
Mouse CD5L Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD5L in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 14 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CD5L Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CD5L Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CD5L biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color