Learn More
Sino Biological Recombinant Anti-CD6 Antibody (PerCP), Rabbit Monoclonal
Supplier:  Sino Biological 50711R021C100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD6 (rM CD6; Catalog#50711-M08H; Q91WN5; Met1-Gly396) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-240-8945