Learn More
Sino Biological Recombinant Anti-CD6 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50711R00220
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD6 / Cluster of Differentiation 6 (rM CD6 / Cluster of Differentiation 6; Catalog#50711-M08H; Q91WN5; Met1-Gly396).
Catalog No. 50-241-2585