Learn More
Sino Biological Recombinant Anti-CD6 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50711R021P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD6 (rM CD6; Catalog#50711-M08H; Q91WN5; Met1-Gly396) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-2586