Learn More
Sino Biological Mouse CD6 Matched ELISA Antibody Pair Set 5 Plates
Supplier: Sino Biological SEK507115
The Mouse CD6 / Cluster of Differentiation 6 ELISA Pair Set is for the quantitative determination of Mouse CD6 / Cluster of Differentiation 6.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse CD6 / Cluster of Differentiation 6 coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse CD6 / Cluster of Differentiation 6 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse CD6 / Cluster of Differentiation 6 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse CD6 / Cluster of Differentiation 6 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.