Learn More
Sino Biological Anti-CD6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50711RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD6 (rM CD6; Catalog#50711-M08H; Q91WN5; Met1-Gly396). CD6 specific IgG was purified by Mouse CD6 affinity chromatography.
Catalog No. 50-240-9842