Learn More
Sino Biological Mouse CD63 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 50557M08H100
Encompass_Preferred
A DNA sequence encoding the Mouse CD63 (NP_031679.1) (Ala103-Ile203) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2861