Learn More
Sino Biological Mouse CD7 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 50393M02H1
Encompass_Preferred
A DNA sequence encoding the Mouse CD7 (NP_033984.1) (Met1-Pro150) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3197