Learn More
Sino Biological Mouse CD70 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 51129M07H100
Encompass_Preferred
A DNA sequence encoding the Mouse CD70 (NP_035747.1) (Gln47-Pro195) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-2451