Learn More
Sino Biological Recombinant Mouse CD70/CD27 Ligand Protein (His Tag), Trimer, HPLC-verified
Supplier:  Sino Biological 51129M07H2100
Encompass_Preferred
A DNA sequence encoding the Mouse CD70 (NP_035747.1) (Gln47-Pro195) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-2985