Learn More
Sino Biological Recombinant Mouse CD70/CD27 Ligand Protein (mFc Tag), HPLC-verified
Supplier:  Sino Biological 51129M04H1100
Encompass_Preferred
A DNA sequence encoding the Mouse CD70 (NP_035747.1) (Gln47-Pro195) was expressed with the Fc region of mouse IgG1 at the N-terminus. This construct has a trimer design.
Catalog No. 50-255-2987