Learn More
Sino Biological Recombinant Mouse CD70/CD27 Ligand Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 51129M49HB20
Encompass_Preferred
A DNA sequence encoding the Mouse CD70 (NP_035747.1) (Gln47-Pro195) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-8684