Learn More
Sino Biological Anti-CD72 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 109330T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse CD72, and purified by antigen affinity chromatography.
Catalog No. 50-255-5057