Learn More
Sino Biological Recombinant Cynomolgus, Rhesus CD39/ENTPD1 Protein (His Tag)
Supplier:  Sino Biological 90310C08B50
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus ENTPD1 (XP_015311944.1 & XP_015003298.1) (Thr60-Val500) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus ENTPD1 sequences are identical.
Catalog No. 50-224-6884