Learn More
Sino Biological Anti-CD79B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50754RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD79B / Ig-beta (rM CD79B / Ig-beta; Catalog#50754-M08H; P15530; Met1-Asp158). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3108