Learn More
ACROBiosystems ACROBiosystems Mouse CD79B Protein, His Tag, low endotoxin (MALS verified)
Supplier:  ACROBiosystems CDBM52H41MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse CD79B, His Tag, low endotoxin (CDB-M52H4) is expressed from human 293 cells (HEK293). It contains AA Val 26 - Asp 158 (Accession # P15530-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.0 kDa. The protein migrates as 28-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7243