Learn More
Sino Biological Mouse CD8 alpha Matched ELISA Antibody Pair Set 15 Plates
Supplier: Sino Biological SEK5038915
The Mouse CD8a ELISA Pair Set is for the quantitative determinationof Mouse CD8a.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD8a coated on a 96-well plate. Standards and samples are added to the wells, and any CD8a present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-CD8a polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of CD8a present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.