Learn More
Apexbio Technology LLC Mouse CD8+ T Cell Isolation Kit (Negative Selection) 100assays(for 1×10⁹ cells)
SDP
Catalog No. 5000093315
Encompass_Preferred
Mouse CD8plus T Cell Isolation Kit Negative Selection is a reagent kit designed for the isolation of untouched CD8 T cells It employs a negative immunomagnetic selection mechanism where non-CD8 T cells are labeled with biotin-conjugated antibodies and subsequently removed using streptavidin-conjugated magnetic beads The kit is optimized for use with murine splenic or lymph node single-cell suspensions The isolated CD8 T cell population typically achieves a purity of over 95% Based on these properties Mouse CD8plus T Cell Isolation Kit Negative Selection is suitable for downstream applications including flow cytometry functional assays or adoptive transfer experiments
Catalog No. 50-000-93315 Supplier Apexbio Technology LLC Supplier No. CS1008100
Mouse CD8plus T Cell Isolation Kit Negative Selection is a reagent kit designed for the isolation of untouched CD8 T cells It employs a negative immunomagnetic selection mechanism where non-CD8 T cells are labeled with biotin-conjugated antibodies and subsequently removed using streptavidin-conjugated magnetic beads The kit is optimized for use with murine splenic or lymph node single-cell suspensions The isolated CD8 T cell population typically achieves a purity of over 95% Based on these properties Mouse CD8plus T Cell Isolation Kit Negative Selection is suitable for downstream applications including flow cytometry functional assays or adoptive transfer experiments