Learn More
Sino Biological Recombinant Anti-CD83 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 50694R134F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD83 / HB15 (rM CD83 / HB15; Catalog#50694-M08H; O88324; Met1-Arg133) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-255-3145