Learn More
Sino Biological Recombinant Anti-CD83 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50694R005100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD83 (rM CD83; Catalog#50694-M08H; O88324; Met1-Arg133).
Catalog No. 50-255-3150