Learn More
Sino Biological Recombinant Anti-CD83 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50694R134P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD83 / HB15 (rM CD83 / HB15; Catalog#50694-M08H; O88324; Met1-Arg133) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-3142