Learn More
Sino Biological Mouse CD83 MAb 25TS (Sino Biological)
Supplier:  Sino Biological 50694R101A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD83 (rM CD83; Catalog#50694-M08H; O88324; Met1-Arg133) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-237-1383