Learn More
Sino Biological Anti-CD83 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50694RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD83 (rM CD83; Catalog#50694-M08H; O88324; Met 1-Arg 133). CD83 specific IgG was purified by Mouse CD83 affinity chromatography.
Catalog No. 50-255-3141