Learn More
Sino Biological Mouse CD86 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50068T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD86/B7-2 (rM CD86/B7-2; Catalog#50068-M08H; NP_062261.3; Met1-Glu245). CD86/B7-2 specific IgG was purified by Mouse CD86/B7-2 affinity chromatography.
Catalog No. 50-237-1749