Learn More
Sino Biological Recombinant Mouse B7-2/CD86 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 50068M02H100
Encompass_Preferred
A DNA sequence encoding the mouse CD86 (NP_062261.3) (Met1-Glu245) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-3495