Learn More
Sino Biological Recombinant Mouse B7-2/CD86 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50068M08HB20
Encompass_Preferred
A DNA sequence encoding the Mouse CD86 (NP_062261.3) (Met1-Glu245) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-3493