Learn More
Sino Biological Recombinant Mouse CD8A & CD8B Heterodimer Protein (Flag & His Tag), HPLC-verified
Supplier:  Sino Biological CT076M2508H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Tyr196) of mouse CD8A (NP_001074579.1) was fused with a flag tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the extracellular domain (Met1-Thr175) of mouse CD8B (NP_033988.1) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the mouse CD8A&CD8B heterodimer was purified.
Catalog No. 50-228-3137