Learn More
Sino Biological Human CD8a / Lyt2 Protein (Fc Tag) 1mg
Supplier:  Sino Biological 10980H02H1
Encompass_Preferred
A DNA sequence encoding the human CD8A (NP_001759.3) (Met1-Asp182) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-222-3368