Learn More
Sino Biological Mouse CD8A PAb 100ul (Sino Biological)
Supplier:  Sino Biological 40905V07E100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD8 alpha (rM CD8 alpha; Catalog#50389-M08H; NP_001074579.1; Met1-Tyr196). CD8 alpha specific IgG was purified by Mouse CD8 alpha affinity chromatography.
Catalog No. 50-237-4321