Learn More
Sino Biological Recombinant Mouse CD8 alpha Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50389M08HB100
Encompass_Preferred
A DNA sequence encoding the Mouse CD8a (NP_001074579.1) (Met1-Tyr196) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-3287