Learn More
Biolegend FITC anti-mouse CD95 (Fas)
Supplier:  Biolegend 152606
Encompass_Preferred
FITC anti-mouse CD95 (Fas) [SA367H8]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1336171