Learn More
Sino Biological Anti-CDC37 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50826T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CDC37 (Catalog#50826-M20B; Q61081; Met1-Ala379). CDC37 specific IgG was purified by Mouse CDC37 affinity chromatography.
Catalog No. 50-255-3065