Learn More
Sino Biological Recombinant Anti-CDCP1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 51110R00120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CDCP1 (rM CDCP1; Catalog#51110-M08H; Q5U462; Met1-Leu666).
Catalog No. 50-255-3005