Learn More
Sino Biological Mouse CDH1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 50671R117100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse E-Cadherin/CDH1/E-cad/CD324 (rM E-Cadherin/CDH1/E-cad/CD324; Catalog#50671-M08H; P09803; Met1-Val709).
Catalog No. 50-237-3998