Learn More
Sino Biological Anti-H Cadherin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51163T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CDH13 / Cadherin-13 / H Cadherin (Catalog#51163-M08H; NP_062681.2; Met1-Ala692). CDH13 / Cadherin-13 / H Cadherin specific IgG was purified by Mouse CDH13 / Cadherin-13 / H Cadherin affinity chromatography.
Catalog No. 50-255-2974