Learn More
Sino Biological Anti-N-Cadherin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 109321T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse N Cadherin, and purified by antigen affinity chromatography.
Catalog No. 50-255-5058