Learn More
Sino Biological Recombinant Anti-VE-Cadherin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50192R228100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse VE-Cadherin / CD144 / CDH5 (rM VE-Cadherin / CD144 / CDH5; Catalog#50192-M08H; NP_033998.2; Met1-Ala592).
Catalog No. 50-240-8137