Learn More
Sino Biological Recombinant Anti-VE-Cadherin Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50192R201P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse VE-Cadherin / CD144 / CDH5 (rM VE-Cadherin / CD144 / CDH5; Catalog#50192-M08H; NP_033998.2; Met1-Ala592) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-8238