Learn More
Sino Biological Mouse CDH5 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 50192R311100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse VE-Cadherin / CD144 / CDH5 (rM VE-Cadherin / CD144 / CDH5; Catalog#50192-M08H; NP_033998.2; Met1-Ala592).
Catalog No. 50-237-2808