Learn More
Sino Biological Anti-VE-Cadherin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50192T56100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse VE-Cadherin/CD144 (rM VE-Cadherin/CD144; Catalog#50192-M08H; NP_033998.2; Met1-Ala592). VE-Cadherin/CD144 specific IgG was purified by Mouse VE-Cadherin/CD144 affinity chromatography.
Catalog No. 50-240-8586